Pesticide standard, Fluquinconazole, reference material, Honeywell Fluka™
HONCR1688-100MG
:
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical Standard
- CAS No.:136426-54-5
- Description:Fluquinconazole
- Formula:C₁₆H₈Cl₂FN₅O
- Storage temperature:Ambient
- Description:Fluquinconazole
- Form:Crystalline
High quality organic reference materials produced according to ISO 9001.
: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.