Order Entry
Finland
ContactUsLinkComponent
Pharmaceutical analytical standard, Norfloxacin-d5, reference material, Honeywell Fluka™
Pharmaceutical analytical standard, Norfloxacin-d5, reference material, Honeywell Fluka™
Catalog # HONCR1654-10MG
CAS Number:  
Pharmaceutical analytical standard, Norfloxacin-d5, reference material, Honeywell Fluka™
Catalog # HONCR1654-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical Standard
  • CAS No.:
    1015856-57-1
  • Description:
    Norfloxacin-d5
  • Formula:
    C₁₆H₁₃D₅FN₃O₃
  • Storage temperature:
    Ambient
  • Description:
    Norfloxacin-d5
  • Form:
    Solid

Specifications

About this item

High quality organic reference materials produced according to ISO 9001.

Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.