Order Entry
Finland
ContactUsLinkComponent
Pesticide standard, Clothianidin, reference material, Honeywell Fluka™
Pesticide standard, Clothianidin, reference material, Honeywell Fluka™
  HONCR1603-100MG
 :  
Pesticide standard, Clothianidin, reference material, Honeywell Fluka™
  HONCR1603-100MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    100 mg
  • Standards:
    Analytical Standard
  • CAS No.:
    210880-92-5
  • Description:
    Clothianidin
  • Formula:
    C₆H₈ClN₅O₂S
  • Storage temperature:
    Ambient
  • Description:
    Clothianidin
  • Form:
    Powder

 

 

High quality organic reference materials produced according to ISO 9001.

 : Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.