Order Entry
Finland
ContactUsLinkComponent
 
Organic analytical standards, Ethylene glycol monobutyl ether, reference material, Honeywell Fluka™
  HONCR1595-5ML
 :  
Organic analytical standards, Ethylene glycol monobutyl ether, reference material, Honeywell Fluka™
  HONCR1595-5ML
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    5 mL
  • Standards:
    Analytical Standard
  • CAS No.:
    111-76-2
  • Description:
    Ethylene glycol monobutyl ether
  • Formula:
    CH₃(CH₂)₃OCH₂CH₂OH
  • Storage temperature:
    Ambient
  • Description:
    Ethylene glycol monobutyl ether
  • Form:
    Liquid

 

 

High quality organic reference materials produced according to ISO 9001.

 : Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.