Order Entry
Finland
ContactUsLinkComponent
Supelco® Organic Reference Standard, PCB No 5
Supelco® Organic Reference Standard, PCB No 5
Catalog # 35588-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, PCB No 5
Catalog # 35588-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    16605-91-7
  • Description:
    PCB No 5
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00055244
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2,3-Dichlorobiphenyl; 2,3-PCB
  • Molecular weight:
    223,10
  • Flash point:
    Not applicable

Specifications

About this item

Caution: P260 - P273 - P314 - P391 - P501