Order Entry
Finland
ContactUsLinkComponent
Pesticide reference standard, Febantel, VETRANAL(TM), analytical standard, Supelco®
Pesticide reference standard, Febantel, VETRANAL(TM), analytical standard, Supelco®
Catalog # 33981-100MG-R
Supplier:  Merck
CAS Number:  
undefined
Pesticide reference standard, Febantel, VETRANAL(TM), analytical standard, Supelco®
Catalog # 33981-100MG-R
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    58306-30-2
  • Description:
    Febantel, VETRANAL(TM)
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₂₀H₂₂N₄O₆S
  • Description:
    Febantel, VETRANAL(TM)

Specifications

About this item

Febantel might be utilised as a source of reference standard in the determination of febantel in samples of animal feed and pharmaceutical formulations, using high performance liquid chromatography (HPLC) and high performance liquid chromatography coupled with tandem mass spectrometry (HPLC-MS-MS).