Order Entry
Finland
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Clodinafop-Propargyl
Supelco® PESTANAL® Organic Reference Standard, Clodinafop-Propargyl
Catalog # 31676-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Clodinafop-Propargyl
Catalog # 31676-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    105512-06-9
  • Description:
    Clodinafop-propargyl
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 + H332 - H373 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD01632328
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2-Propynyl (R)-2-[4-(5-chloro-3-fluoro-2-pyridinyloxy)phenoxy]propionate
  • Formula:
    C₁₇H₁₃ClFNO₄
  • Molecular weight:
    349,74
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS08, GHS07, GHS09
Caution: P260 - P264 - P273 - P301 + P312 - P304 + P340 + P312 - P314