Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Environmental Test Kits
CalVer B concentrate and water
CalVer B concentrate and water
CalVer B concentrate and water
Catalog # HACH2190304
Supplier:
Hach
CAS Number:
Expand image
CalVer B concentrate and water
Catalog # HACH2190304
Supplier:
Hach
CAS Number:
Specifications
Pk:
4
Description:
CalVer B concentrate and water
Sample size:
946 ml
Specifications
About this item
Product Details
Product Details