Hemin chloride (from porcine), Calbiochem®, Millipore®
3741-5
:
- Pack type:Plastic ampoule
- Pk:5 g
- Seq:3173
- Purity:Homogeneous by TLC
:
:651.95 g/mol
:14,04644
:M-Clarity™ quality level = MQ100
:Ambient
:240-140-1
:16009-13-5
:C₃₄H₃₂ClFeN₄O₄
:MFCD00010726