Order Entry
Finland
ContactUsLinkComponent
Test strips, tumeric paper for qualitative detection of borates
Test strips, tumeric paper for qualitative detection of borates
Catalog # MANA90747
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test strips, tumeric paper for qualitative detection of borates
Catalog # MANA90747
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Pk:
    200
  • Description:
    Boric acid and borates H₃BO₃, BO₃³⁻
  • Range:
    20 mg/l B / 100 mg/l H₃BO₃

Specifications

About this item

The test paper allows the quick and easy detection of boric acid and borates. It is impregnated with curcumin, the yellow dye extracted from the roots of Curcuma tinctoria (yellow ginger).

  • Colour changes: Yellow to red

The paper is dipped into the sample containing hydrochloric acid (pH 1 to 2). Afterwards it is allowed to dry.

In presence of borate the paper turns orange to red. Dipping into conc. sodium hydroxide causes a green-black colour. In the absence of borate, sodium hydroxide leads to a brown-red colour.

Packaging: Pack of 200 strips 20x70 mm.