Methyl blue microbiology stain (C.I. 42780)
Catalog # 34015.182
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Identification:Passes test
Specification Test Results
About this item
:
:799.81 g/mol
CAS Number:28983-56-4
Formula:C₃₇H₂₇N₃Na₂O₉S₃
MDL Number:MFCD00058509