Pharmaceutical analytical standard, Deracoxib-d3, reference material, Honeywell Fluka™
Catalog # HONCR1466-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:2012598-48-8
- Description:Deracoxib-d₃
- Formula:C₁₇H₁₁D₃F₃N₃O₃S
- Storage temperature:Ambient
- Description:Deracoxib-d₃
- Freezing point:157...158 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results