Pharmaceutical analytical standard, Piroxicam, reference material, Honeywell Fluka™
HONCR1458-10MG
:
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:36322-90-4
- Description:Piroxicam
- Formula:C₁₅H₁₃N₃O₄S
- Storage temperature:Ambient
- Description:Piroxicam
- Freezing point:198...200 °C
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results