Order Entry
Finland
ContactUsLinkComponent
Pharmaceutical analytical standard, Isopropylaminoantipyrine-d6, reference material, Honeywell Fluka™
Pharmaceutical analytical standard, Isopropylaminoantipyrine-d6, reference material, Honeywell Fluka™
Catalog # HONCR1453-10MG
CAS Number:  
Pharmaceutical analytical standard, Isopropylaminoantipyrine-d6, reference material, Honeywell Fluka™
Catalog # HONCR1453-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    1330180-51-2
  • Description:
    Isopropylaminoantipyrine-d₆
  • Formula:
    C₁₄H₁₃D₆N₃O
  • Storage temperature:
    Ambient
  • Description:
    Isopropylaminoantipyrine-d₆
  • Freezing point:
    79 °C

Specifications

About this item

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results