Pharmaceutical analytical standard, Salmeterol-13C6 xinafoate, reference material, Honeywell Fluka™
Catalog # HONCR1416-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- Description:Salmeterol-¹³C₆ xinafoate
- Formula:¹³C6C₁₉H₃₇NO₄ · C₁₁H₈O₃
- Storage temperature:Ambient
- Description:Salmeterol-¹³C₆ xinafoate
- Freezing point:116...118 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results