Order Entry
Finland
ContactUsLinkComponent
 
Organic analytical standards, Butyl 4-hydroxybenzoate-ring-13C6 solution, 50 μg/mL in acetone, reference material, Honeywell Fluka™
Catalog # HONCR1349-1ML
CAS Number:  
Organic analytical standards, Butyl 4-hydroxybenzoate-ring-13C6 solution, 50 μg/mL in acetone, reference material, Honeywell Fluka™
Catalog # HONCR1349-1ML
CAS Number:  

Specifications

  • Concentration:
    50 μg/ml in acetone
  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    1416711-53-9
  • Description:
    Butyl 4-hydroxybenzoate-ring-¹³C₆ solution
  • Formula:
    ¹³C₆C₅H₁₄O₃
  • Storage temperature:
    –20 °C
  • Description:
    Butyl 4-hydroxybenzoate-ring-¹³C₆ solution
  • Flash point:
    –17 °C
  • Boiling point:
    55...57 °C
  • Freezing point:
    –95 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results