Organic analytical standards, Propyl 4-hydroxybenzoate-ring-13C6 solution, 50 μg/ml in acetone, reference material, Honeywell Fluka™
Catalog # HONCR1348-1ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Concentration:50 μg/ml in acetone
- Pack type:Glass bottle
- Pk:1 mL
- Description:Propyl 4-hydroxybenzoate-ring-¹³C₆ solution
- Formula:¹³C₆C₄H₁₂O₃
- Storage temperature:–20 °C
- Description:Propyl 4-hydroxybenzoate-ring-¹³C₆ solution
- Flash point:–17 °C
- Boiling point:55...57 °C
- Freezing point:–95 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results