Pesticide reference standard, Methasulfocarb, neat, Honeywell Fluka™
Catalog # HONCR1149-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mg
- CAS No.:66952-49-6
- Description:Methasulfocarb
- Formula:C₉H₁₁NO₄S₂
- Storage temperature:Ambient
- Description:Methasulfocarb
- Freezing point:138 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results