Order Entry
Finland
ContactUsLinkComponent
 
Pesticide reference standard, 2-(Thiocyanatomethylthio)benzothiazole, neat, Honeywell Fluka™
  HONCR1081-25MG
 :  
Pesticide reference standard, 2-(Thiocyanatomethylthio)benzothiazole, neat, Honeywell Fluka™
  HONCR1081-25MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mg
  • CAS No.:
    21564-17-0
  • Description:
    2-(Thiocyanatomethylthio)benzothiazole
  • Formula:
    C₉H₆N₂S₃
  • Storage temperature:
    Ambient
  • Description:
    2-(Thiocyanatomethylthio)benzothiazole
  • Flash point:
    189 °C
  • Boiling point:
    191 °C

 

 

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results