Industrial gloves, Camapur® Comfort 616
Supplier: KCL by Honeywell
Certificates
About this item
These seamless polyamide liners feature a PU palm coating which provides excellent grip when handling greasy and oily parts.
- Excellent fit
- Very breathable
- Good mechanical resistance
- Allows working without user fatigue
- Less sensitive to soiling
- Washable
Ideal for usage in the electronics and automotive industry, assembly works, for precision mechanics, protection of products, metal and plastics processing and for storage and commission work.
Certifications: EN 388 (3131); Cat. II
Specifications
- Catalog No:
- KCHL616.8
- KCHL616.7
- KCHL616.11
- KCHL616.6
- KCHL616.10
- KCHL616.9
- Model:
- Comfort 616
- Comfort 616
- Comfort 616
- Comfort 616
- Comfort 616
- Comfort 616
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Polyamide
- Polyamide
- Polyamide
- Polyamide
- Polyamide
- Polyamide
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388 (3131)
- EN 388 (3131)
- EN 388 (3131)
- EN 388 (3131)
- EN 388 (3131)
- EN 388 (3131)