Order Entry
Finland
ContactUsLinkComponent
UST MIX, certified reference material, Supelco®
UST MIX, certified reference material, Supelco®
Catalog # UST111-1.5ML
Supplier:  Merck
CAS Number:  
UST MIX, certified reference material, Supelco®
Catalog # UST111-1.5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml in methanol
  • Pk:
    1,5 mL
  • Description:
    UST mix

Specifications

About this item