Order Entry
Finland
ContactUsLinkComponent
Hydrochloric acid 4.0 M HCl (4N) volumetric solution, Fluka™
Irritant
Corrosive
Hydrochloric acid 4.0 M HCl (4N) volumetric solution, Fluka™
Catalog # 35402-10L
CAS Number:  
Irritant
Corrosive
Hydrochloric acid 4.0 M HCl (4N) volumetric solution, Fluka™
Catalog # 35402-10L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 L
  • Seq:
    5214

Specifications

Specification Test Results

  • Vapor Pressure:
    190 mmHg ( 25 °C)
  • Factor:
    0.998 - 1.002
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
Density:1.072 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1789
ADR:8,II
 :36.46 g/mol
 :GHS07
 :GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324