Order Entry
Finland
ContactUsLinkComponent
Gauze bandage, Peha®
Gauze bandage, Peha®
Catalog # HART3040530
Supplier:  HARTMANN PAUL
Gauze bandage, Peha®
Catalog # HART3040530
Supplier:  HARTMANN PAUL

Specifications

  • Description:
    Gauze bandage, Peha®, 1 folding box with 20 rolls
  • L×W:
    4 m × 100 mm
  • Pk:
    20

Specifications

About this item

Gauze bandage made of non-elastic fabric (60% viscose/40% cotton).

  • With woven edges
  • Sold as rolls

Ideal for use as wound dressings or padding material. Also suitable as a carrier fabric for zinc glue dressings as well as first aid emergency bandages.