Order Entry
Finland
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, Triazine Pesticides Standards Mix
Supelco® TraceCERT® Organic Reference Standard, Triazine Pesticides Standards Mix
Catalog # CRM48392
Supplier:  Merck
CAS Number:  
Supelco® TraceCERT® Organic Reference Standard, Triazine Pesticides Standards Mix
Catalog # CRM48392
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 μg/ml each component in methanol
  • Pk:
    1 mL
  • Standards:
    Certified reference material, TraceCERT®
  • Description:
    Triazine pesticides standards mix
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370 - H412
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Flash point:
    9,7 °C - Closed cup

Specifications

About this item

  • GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P273 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311