Order Entry
Finland
ContactUsLinkComponent
Supelco® Organic Reference Standard, MDI-DBA-d₉ Oligomers Standard
Supelco® Organic Reference Standard, MDI-DBA-d₉ Oligomers Standard
Catalog # CRM40604
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, MDI-DBA-d₉ Oligomers Standard
Catalog # CRM40604
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    In methanol (varied conc.)
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • Description:
    MDI-DBA-d₉ oligomers standard
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370
  • Shipping conditions:
    WICE
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Methylenediphenyl diisocyanate dibutylamine-d₉; p-MDI; polymeric-MDI
  • Flash point:
    9,7 °C - Closed cup
  • Pk Info:
    Ampule of 1 ml

Specifications

About this item

  • GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311