Order Entry
Finland
Orders LinkContactUsLinkComponent
Antifade solution

Antifade solution

2×0.5 ml, 1 mL
Catalog # ABNOU0032
Supplier:   Abnova
Antifade solution
Catalog # ABNOU0032
Supplier:  Abnova
CAS Number:  
Antifade solution

2×0.5 ml, 1 mL

CAS Number:  
Price
196,00 €
Log in for availability
1 * 1 mL
Product Details & Documents

Antifade Solution is comprised of Antifade and glycerol in an aqueous buffer solution.

  • for Fluorescent In Situ Hybridization (Cell)
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pk:
    1 mL
  • Pk Info:
    2×0.5 ml

Specifications