Supelco® Organic Reference Standard, Omeprazole Related Compound B
Catalog # PHR1986-30MG
Supplier: Merck
CAS Number:
Specifications
- Pk:30 mg
- Standards:Pharmaceutical Secondary Standard; Certified Reference Material
- CAS No.:37052-78-1
- Description:Omeprazole related compound B
- Storage conditions:2…8 °C
- Hazard class description:H315 - H319 - H335
- Shipping conditions:WICE
- MDL No.:MFCD00134581
- M-Clarity quality level:MQ300
- Synonym(s):5-Methoxy-2-benzimidazolethiol; 5-METHOXY-2- BENZIMIDAZOLETHIOL
- Melting point:261 - 263 °C (lit.)
- Molecular weight:180,23
Specifications
About this item
- GHS signal: GHS07
Caution: P305 + P351 + P338