Supelco® Organic Reference Standard, beta-Pinene solution
Catalog # CRM40433
Supplier: Merck
CAS Number:
Specifications
- Concentration:2000 µg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:127-91-3
- Description:beta-Pinene solution
- Storage conditions:2…8 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:WICE
- M-Clarity quality level:MQ300
- Flash point:9,7 °C - Closed cup
- Pk Info:Ampule of
Specifications
About this item
- GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311