Order Entry
Finland
ContactUsLinkComponent
6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE) 90 mg/mL in anhydrous DMSO amine-reactive fluorescent dye
6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE) 90 mg/mL in anhydrous DMSO amine-reactive fluorescent dye
Catalog # 90097.
Supplier:  Biotium
CAS Number:  
undefined
6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE) 90 mg/mL in anhydrous DMSO amine-reactive fluorescent dye
Catalog # 90097.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    100 µl
  • Seq:
    1286

Specifications

About this item

 :
 :527.52 g/mol
Storage Temperature:Freezer
CAS Number:150810-69-8
Formula:C₂₉H₂₅N₃O₇