Order Entry
Finland
ContactUsLinkComponent
Diaphragm pumps, VARIO® select
Diaphragm pumps, VARIO® select
Supplier:  VACUUBRAND
Certificates
undefined
Diaphragm pumps, VARIO® select
Supplier:  VACUUBRAND
Specifications
  • Catalog No:
  • 131-0050
  • 131-0033
  • Model:
  • MV 2 VARIO select (CEE-plug)
  • MD 4 VARIO select (CEE-plug)
  • No. of stages:
  • 4
  • 3
  • Flow rate:
  • 55 l/min
  • 95 l/min
  • Ultimate vacuum:
  • 0,3 mbar (0,2 torr)
  • 1,0 mbar (0,75 torr)
Specifications