Order Entry
Finland
ContactUsLinkComponent
Hygiene kit for earmuffs, Peltor™
Hygiene kit for earmuffs, Peltor™
Catalog # MMMAHY79
Supplier:  3M
undefined
Hygiene kit for earmuffs, Peltor™
Catalog # MMMAHY79
Supplier:  3M
Specifications
  • Description:
    Hygiene kit for earmuffs Peltor™ ProTac, Tactical™ XP, FM-Radio, Alert™ and LiteCom™
  • Pk:
    1
Specifications
About this item
These hygiene kits include replacement ear cushions, foam liners and rings. For use with variety of Peltor™ products, including ProTac, Tactical™ XP, FM-Radio, Alert™ and LiteCom™.
  • Maintain hygiene of communications headsets that see heavy use or multiple users
  • Replace at least twice a year to ensure constant attenuation, hygiene and comfort
Colour: Black