Chemical resistant gloves, uvex rubiflex S NB27B
Supplier: UVEX
Certificates
About this item
These lightweight stockinette gloves feature a special full NBR coating and offer a high level of protection against a diverse range of chemical and mechanical risks. With outstanding tactile feel, touch monitors can also be operated while wearing the glove.
- Outstanding wearer comfort and ergonomic fit
- Good resistance to grease, mineral oils and many chemicals
- Extremely high flexibility
- Good grip in damp and wet areas
Areas of application: Automotive and chemical industry, laboratories, painting work and food processing.
Certifications: EN 388:2016 (2111X), EN ISO 374-1:2016 Type A (JKNOPT)
Food contact approved
OEKO-TEX® Standard 100
Food contact approved
OEKO-TEX® Standard 100
Specifications
- Catalog No:
- UVEX6027108
- UVEX6027109
- UVEX6027111
- UVEX6027110
- Model:
- NB27B
- NB27B
- NB27B
- NB27B
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- EN norm:
- EN 388 (2111X), EN ISO 374-1:2016 Type A (JKNO PT)
- EN 388 (2111X), EN ISO 374-1:2016 Type A (JKNO PT)
- EN 388 (2111X), EN ISO 374-1:2016 Type A (JKNO PT)
- EN 388 (2111X), EN ISO 374-1:2016 Type A (JKNO PT)