Wall Bracket for 30 L Bio-bin® Boxes
Catalog # ECONEXBB30WB
Supplier: ECONIX LTD
Specifications
- Description:Wall bracket, chrome plated, for 30 l Bio-bin® boxes
- Pk:5
Specifications
About this item
This wall bracket is used in areas where the bin needs to be kept off the floor.
- Easy to attach to a wall or to the side of a trolley
- Chrome plated for easy cleaning
- Fixed into place with M8 screws and plugs depending on the wall type (not supplied)
Ordering information: Supplied without screws.