Cut resistant gloves, uvex unidur 6648
Supplier: UVEX
Certificates
About this item
These HPPE/elastane cut protection gloves with PU coating on palm and fingertips provide a cut protection level of 3/B. They are extremely sensitive and exceptionally well suited to fine assembly work in which a high level of tactility and cut protection are required.
- Outstanding mechanical abrasion resistance
- Good grip in dry and slightly damp areas
- Good tactile feel, high flexibility and good wearer comfort
Areas of application: Automotive industry, repair work, light to medium metal processing and packaging.
Certifications: EN 388:2016 (4342B).
Specifications
- Catalog No:
- UVEX6093207
- UVEX6093206
- UVEX6093210
- UVEX6093209
- UVEX6093208
- UVEX6093211
- Model:
- 6648
- 6648
- 6648
- 6648
- 6648
- 6648
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- HPPE
- HPPE
- HPPE
- HPPE
- HPPE
- HPPE
- Finish:
- PU coating on palm and fingertips
- PU coating on palm and fingertips
- PU coating on palm and fingertips
- PU coating on palm and fingertips
- PU coating on palm and fingertips
- PU coating on palm and fingertips
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)