Order Entry
Finland
ContactUsLinkComponent
Biological indicators, traditional spore strips
Biological indicators, traditional spore strips
Catalog # NAMSBS52306
Supplier:  CROSSTEX
Biological indicators, traditional spore strips
Catalog # NAMSBS52306
Supplier:  CROSSTEX

Specifications

  • Description:
    Bacillus subtilis spore strips, cell line 5230, 10⁶ spores/strip
  • Pk:
    100

Specifications

About this item

Bacillus subtilis strips for monitoring low temperature steam sterilisation processes.

  • Inoculated filter paper
  • Convenient shelf packages of 100 units per box

The strips can be easily removed from the glassine pouch by tearing or peeling the pouch open for transfer to culture media or challenge device assembly.

Shelf life: 15 months

Room temperature storage conditions (15 to 30 °C), relative humidity of 30% to 80% is recommended.

Delivery: Supplied with a Certificate of Analysis.
Packaging: Individually packaged in easy-to-handle glassine pouches.