Order Entry
Finland
ContactUsLinkComponent
Autoclavable cleanroom goggles, 611
Autoclavable cleanroom goggles, 611
Catalog # 111-2515
Supplier:  UNIVET
undefined
Autoclavable cleanroom goggles, 611
Catalog # 111-2515
Supplier:  UNIVET

Specifications

  • Type:
    Anti-UV, anti-fog, autoclavable, directly vented
  • Colour:
    Light blue
  • Lenses:
    Clear PC
  • Pk:
    5

Specifications

About this item

These 611 cleanroom goggles are sterilisable in an autoclave with steam and also with radiation.

  • Panoramic goggles with ventilation
  • Frame and lens made of anallerginic material
  • Polycarbonate lens (2 mm)
  • Extra-wide elastic strap UNVT3PA299 is sold separately

Weight: 104 g

Certifications: EN 166