Order Entry
Finland
ContactUsLinkComponent
Coleman® liquid fuel
Coleman® liquid fuel
Catalog # CAMP2000016589
Supplier:  CAMPING GAZ
CAS Number:  
Coleman® liquid fuel
Catalog # CAMP2000016589
Supplier:  CAMPING GAZ
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :GHS02
Storage Temperature:Ambient