Order Entry
Finland
ContactUsLinkComponent
Disinfectant, ClearKlens Tego 2000 SC, VH25
Disinfectant, ClearKlens Tego 2000 SC, VH25
  DIVY100868202
 :  Diversey
undefined
Disinfectant, ClearKlens Tego 2000 SC, VH25
  DIVY100868202
 :  Diversey
  • Description:
    ClearKlens Tego 2000 SC disinfectant, VH25
  • Pk:
    50 Bottle
  • pH value:
    8,0 (20 °C)
ClearKlens Tego 2000 SC is a concentrated disinfectant for the application in the pharmaceutical and medical industries.
  • Sterile, amphoteric disinfectant
  • Detex label for proof of irradiation
  • Triple packed to facilitate transport to the clean room
  • No discolouration, low toxicity and odourless
  • Broad spectrum of efficacy
  • Effective in hard and soft water
ClearKlen's Tego 2000 SC is 0,2 μm filtered and triple packaged, validated gamma irradiation process. It is suitable for spray and nebulisation applications.
 : Use biocides safely. Always read the label and product information before use.