DURAN® PURE GLS 80® Bottles
Supplier: DWK Life Sciences
Certificates
About this item
DURAN® PURE bottles clear or amber, with GLS 80 thread and dust cover. DURAN PURE is a range of primary packaging products dedicated to the specific requirements and needs of the pharmaceutical industry.
- Made from DURAN® borosilicate glass 3.3 Type I
- Wide mouth allows easy filling and emptying of bottles
Suitable for manufacturing of pharmaceutical products, storage of bulk drug intermediates and ingredients and transportation of API.
Certifications: The bottles and closures in the range fully meet the requirements laid down in the international pharmacopoeias, and offer full protection for sensitive and valuable pharmaceutical products. DURAN PURE products are available with a full range of documents and certification that meet the detailed requirements of the regulatory authorities.