Perchloric acid ≥70% ACS, Fluka™
:7601-90-3
:
:HClO₄
:MFCD00011325
:100.46 g/mol
:203 °C
:–18 °C
:1.66 g/cm³ (20 °C)
:1873
:5.1,I
:GHS08
:GHS05
:GHS07
:GHS03
:Ambient
- Assay (alkalimetric):69.0 - 72.0 %
- Residue on ignition:Max. 0.003 %
- Iron (Fe):Max. 0.0001 %
- Heavy metals (as Pb):Max. 0.0001 %
- Chloride (Cl):Max. 0.001 %
- Total N:Max. 0.001 %
- Phosphate, silicate (as SiO₂):Max. 0.0005 %
- Sulfate (SO₄):Max. 0.001 %
- APHA:Max. 10