

Calcium oxide 96-100.5% (excluding ignited substances), fine powder, Puriss., meets analytical specification of FCC, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:1305-78-8
:
Formula:CaO
MDL Number:MFCD00010911
:56.08 g/mol
Merck Index:13,01691
Boiling Pt:2.850 °C (1.013 hPa)
Melting Pt:2.572 °C (1.013 hPa)
Density:3.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-138-9
:GHS07
:GHS05
Specification Test Results
- Assay (excluding ignited substance):96 - 100.5 %
- Insoluble in HCl:Max. 0.5 %
- Loss on ignition:Max. 3.5 %
- Arsenic (As):Max. 0.0003 %
- Lead (Pb):Max. 0.0002 %
- Fluoride (F):Max. 0.015 %
- Mg and alkali salts (SO₄):Max. 2.5 %
- Form:powder (fine)
Specification Test Results
Frequently Bought Together