Order Entry
Finland
ContactUsLinkComponent
 
 
Potassium thiocyanate ≥99%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
 
 
Potassium thiocyanate ≥99%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
 

 

  • Assay:
    Min. 99 %
  • Water insoluble matter:
    Max. 0.005 %
  • Insoluble in Ethanol:
    Max. 0.01 %
  • pH (5 %, 25 °C):
    5.3 - 8.5
  • Ammonium (NH₄):
    Max. 0.001 %
  • Copper (Cu):
    Max. 0.0002 %
  • Iron (Fe):
    Max. 1 ppm
  • Sodium (Na):
    Max. 0.005 %
  • Lead (Pb):
    Max. 0.0002 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 0.005 %
  • Sulfate (SO₄):
    Max. 0.001 %
  • Sulfide (S):
    Max. 0.001 %
  • Matter consuming I (as I):
    Max. 0.025 %