Safety needles, Sterican®
Supplier: B. Braun
Certificates
About this item
This single-use needle with safety protection is compatible with all standard Luer lock and Luer slip syringes.
- Made of stainless chromium-nickel steel
- Needle hub made of PP
The safety feature of Sterican® safety needle is in compliance with the EU Directive and provides an intuitive, one-handed activation. It securely and irreversibly locks the needle within the integrated shield for a safe disposal.
Certifications: In compliance with EU Directive 2010/32/EU. Colour coding according to ISO 6009.