HPLC columns, NUCLEOSHELL® PFP
NUCLEOSHELL® PFP based on core-shell particle technology for fast and efficient HPLC.
- Efficient core-shell silica gel
- Pentafluorophenyl propyl (PFP) modification
- Hydrophobic phase with alternative selectivity in comparison to classical C18 modifications
The separation principle based on four retention mechanisms such as polar interactions, dipole-dipole interactions, π-π interactions, hydrophobic interactions.
Suitable for the separation of polar compounds like pharmaceuticals such as antibiotics, aromatic and unsaturated compounds, phenols, halogenated hydrocarbons and isomers. It shows a high retention of basic compounds.
EC guard columns are used for analytical EC columns with the column protection system guard column holder (554-5851).
- Catalog No:
- MANA763536.46
- MANA763538.30
- MANA763534.46
- MANA763532.40
- MANA763532.46
- MANA763534.30
- MANA763534.40
- MANA763536.40
- MANA763534.20
- MANA763538.20
- MANA763532.20
- MANA763532.30
- MANA763536.20
- MANA763536.30
- Carbon load:
- ~3,0%
- ~3,0%
- ~3,0%
- ~3,0%
- ~3,0%
- ~3,0%
- ~3,0%
- ~3,0%
- ~3,0%
- ~3,0%
- ~3,0%
- ~3,0%
- ~3,0%
- ~3,0%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9