Order Entry
Finland
ContactUsLinkComponent
Analytical standard, Hydramethylnon, reference material, Honeywell Fluka™
Analytical standard, Hydramethylnon, reference material, Honeywell Fluka™
Catalog # HONCR1376-100MG
CAS Number:  
Analytical standard, Hydramethylnon, reference material, Honeywell Fluka™
Catalog # HONCR1376-100MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    100 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    67485-29-4
  • Description:
    Hydramethylnon
  • Melting point:
    189…191 °C
  • Formula:
    C₂₅H₂₄F₆N₄
  • Storage temperature:
    Ambient
  • Description:
    Hydramethylnon

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results