Analytical standard, Oxaziclomefon, reference material, Honeywell Fluka™
Catalog # HONCR1285-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:153197-14-9
- Description:Oxaziclomefon
- Melting point:147…152 °C
- Formula:C₂₀H₁₉Cl₂NO₂
- Storage temperature:2…8 °C
- Description:Oxaziclomefon
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results