Analytical standard, Valnemulin hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1264-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:25 mg
- Standards:Analytical standard, neat
- CAS No.:133868-46-9
- Description:Valnemulin hydrochloride
- Melting point:160 °C
- Formula:C₃₁H₅₂N₂O₅S · HCl
- Storage temperature:–20°C
- Description:Valnemulin hydrochloride
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results