Analytical standard, (±)-Naproxen-(methoxy-d₃), reference material, Honeywell Fluka™
Catalog # HONCR1261-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:958293-79-3
- Description:(±)-Naproxen-(methoxy-d₃)
- Melting point:154…159 °C
- Formula:C₁₄D₃H₁₁O₃
- Storage temperature:Ambient
- Description:(±)-Naproxen-(methoxy-d₃)
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results