Analytical standard, Fenthion oxon sulfoxide, reference material, Honeywell Fluka™
HONCR1091-25MG
:
- Pack type:Glass Bottle
- Pk:25 mg
- Standards:Analytical standard, neat
- CAS No.:6552-13-2
- Description:Fenthion oxon sulfoxide
- Melting point:62…66 °C
- Formula:C₁₀H₁₅O₅PS
- Storage temperature:2…8 °C
- Description:Fenthion oxon sulfoxide
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results