Order Entry
Finland
ContactUsLinkComponent
Analytical standard, Etofenprox-(ethoxy-d₅), reference material, Honeywell Fluka™
Analytical standard, Etofenprox-(ethoxy-d₅), reference material, Honeywell Fluka™
  HONCR1048-10MG
 :  
Analytical standard, Etofenprox-(ethoxy-d₅), reference material, Honeywell Fluka™
  HONCR1048-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1705649-55-3
  • Description:
    Etofenprox-(ethoxy-d₅)
  • Formula:
    C₂₅D₅H₂₃O₃
  • Storage temperature:
    Ambient
  • Description:
    Etofenprox-(ethoxy-d₅)

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results